Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181850.2 | NP_862898.1 | 4 - 5 | 2759 | 86 | |
GenBank mRNA | BC060060.1 | - | 2 - 3 | 1117 | 86 | |
BC111891.1 | - | 4 - 5 | 2577 | 40061004 | 86 | |
BC113170.1 | - | 4 - 5 | 2577 | 40061005 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |