Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199147.1 | NP_001186076.1 | 10 - 11 | 1456 | 84 | |
NM_153410.5 | NP_700459.2 | 10 - 11 | 1387 | 84 | ||
GenBank mRNA | AK048691.1 | - | 10 - 11 | 1381 | 84 | |
AK082985.1 | - | 10 - 11 | 1331 | 84 | ||
BC027004.1 | - | 7 - 8 | 738 | 84 | ||
BC071197.1 | - | 10 - 11 | 1453 | 6465025 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |