Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbpj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080927.2 | NP_001074396.1 | 5 - 6 | 716 | 82 | |
NM_001080928.1 | NP_001074397.1 | 5 - 6 | 602 | 82 | ||
NM_001277116.1 | NP_001264045.1 | 7 - 8 | 798 | 82 | ||
NM_009035.5 | NP_033061.3 | 5 - 6 | 618 | 82 | ||
GenBank mRNA | AK028363.1 | - | 7 - 8 | 776 | 82 | |
AK080359.1 | - | 5 - 6 | 511 | 82 | ||
AK090006.1 | - | 5 - 6 | 504 | 82 | ||
AK132145.1 | - | 5 - 6 | 560 | 82 | ||
AK139087.1 | - | 5 - 6 | 503 | 82 | ||
AK152746.1 | - | 5 - 6 | 618 | 82 | ||
AK153163.1 | - | 6 - 7 | 506 | 82 | ||
BC051387.1 | - | 5 - 6 | 547 | 4460290 | 82 | |
S63463.1 | - | 5 - 6 | 619 | 82 | ||
X17459.1 | - | 5 - 6 | 614 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |