Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gast | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010257.3 | NP_034387.3 | 2 - 3 | 256 | 111 | |
GenBank mRNA | AK008062.1 | - | 2 - 3 | 257 | 111 | |
AK008159.1 | - | 2 - 3 | 257 | 111 | ||
AK008313.1 | - | 2 - 3 | 257 | 111 | ||
AK008420.1 | - | 2 - 3 | 257 | 111 | ||
AK008494.1 | - | 2 - 3 | 257 | 111 | ||
AK008649.1 | - | 2 - 3 | 260 | 111 | ||
BC064791.1 | - | 2 - 3 | 257 | 6431366 | 111 | |
X94758.1 | - | 2 - 3 | 261 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons |