Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24613352 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24613353 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24613355 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24613354 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2016981 |
CD775268.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2016981 |
AK145558.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2016981 |
AY335195.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2016981 |
BC018355.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2016981 |
NM_001170591.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1977589 |
CD775268.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1977589 |
AK003730.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1977589 |
AK145558.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1977589 |
AY335195.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1977589 |
BC018355.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
112934 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
512452 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
453887 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
116230 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
376476 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
105777 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
352161 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
55562 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
54609 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
898302 |
AB037665.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
898302 |
AK002659.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
898302 |
AK010277.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
898302 |
AK010518.2 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
898302 |
AK012391.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
898302 |
AK151517.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
898302 |
BC034339.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
898302 |
BC047046.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
898302 |
BC055346.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
898302 |
BC081447.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
898302 |
BC094258.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
898302 |
BC094644.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
898302 |
NM_001048057.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
898302 |
NM_001048058.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
898302 |
NM_023372.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
898302 |
XM_890220.3 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
898302 |
XM_914557.3 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
898302 |
XM_973154.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
898302 |
XM_976669.2 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
898302 |
XM_978287.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
898302 |
XM_984165.2 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
10797 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
111529 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P133143 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P170244 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2016981 |
CD775268.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2016981 |
AK145558.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2016981 |
AY335195.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2016981 |
BC018355.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2016981 |
NM_001170591.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1977589 |
CD775268.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1977589 |
AK003730.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1977589 |
AK145558.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1977589 |
AY335195.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1977589 |
BC018355.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P347728 |
CD775268.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P347728 |
AK003730.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P347728 |
AK145558.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P347728 |
AY335195.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P347728 |
BC018355.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24613351 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24613370 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24613344 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24613346 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24613353 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24613368 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24613356 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24613348 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24613366 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24613363 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24613365 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24613359 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24613360 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24613354 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24613361 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24613357 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05970418 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P133143 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P170244 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P368629 |
AK081950.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P347728 |
CD775268.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P347728 |
AK003730.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P347728 |
AK145558.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P347728 |
AY335195.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P347728 |
BC018355.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P140462 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P135849 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P133143 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P170244 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05437575 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24613352 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24613354 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24613355 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24613353 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613490 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613466 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613375 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613489 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613493 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613499 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613460 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613434 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613404 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613349 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613406 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613340 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613446 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613410 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613399 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613492 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613389 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613408 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613463 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613452 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613364 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613456 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613357 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613425 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613454 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613449 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613359 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613388 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613405 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613373 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613390 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613338 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613472 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613418 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613430 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613435 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613437 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613396 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613336 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613353 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613433 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613428 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613494 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613497 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613381 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613368 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613498 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613391 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613501 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613477 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613427 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613348 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613444 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613413 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613341 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613478 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613432 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613411 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613382 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613384 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613363 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613360 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613352 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613378 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613356 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613400 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613366 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613387 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613442 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613394 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613464 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613377 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613458 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613424 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613441 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24613395 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05018674 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05323918 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613378 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613360 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05754434 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613464 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613366 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613373 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05437577 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613353 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613394 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613354 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613363 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613344 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05018676 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05430492 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613375 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613470 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05430491 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613361 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613466 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613460 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05410510 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613474 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05083451 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613377 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05599245 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05323919 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613372 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05305582 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613395 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613400 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05503577 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613399 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05786745 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05284462 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05410514 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613472 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05431287 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05306109 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613356 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05035481 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613387 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613463 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613336 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613346 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613340 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613475 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613396 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24613351 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05720137 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05306111 |
- |
- |