Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007742.3 | NP_031768.2 | 11 - 12 | 864 | 64 | |
GenBank mRNA | AK132180.1 | - | 11 - 12 | 880 | 64 | |
AK158011.1 | - | 11 - 12 | 881 | 64 | ||
AK159283.1 | - | 11 - 12 | 882 | 64 | ||
AK159285.1 | - | 11 - 12 | 884 | 64 | ||
AK178289.1 | - | 2 - 3 | 85 | 64 | ||
BC042642.1 | - | 4 - 5 | 434 | 64 | ||
BC050014.1 | - | 11 - 12 | 864 | 4225038 | 64 | |
BC059281.1 | - | 11 - 12 | 874 | 3992363 | 64 | |
U08020.1 | - | 11 - 12 | 765 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |