Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007742.3 | NP_031768.2 | 19 - 20 | 1366 | 143 | |
GenBank mRNA | AK132180.1 | - | 19 - 20 | 1385 | 143 | |
AK158011.1 | - | 19 - 20 | 1383 | 143 | ||
AK159283.1 | - | 19 - 20 | 1384 | 143 | ||
AK159285.1 | - | 19 - 20 | 1386 | 143 | ||
BC042642.1 | - | 12 - 13 | 936 | 143 | ||
BC050014.1 | - | 19 - 20 | 1366 | 4225038 | 143 | |
BC059281.1 | - | 19 - 20 | 1376 | 3992363 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |