Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007742.3 | NP_031768.2 | 47 - 48 | 3597 | 103 | |
GenBank mRNA | AK132180.1 | - | 47 - 48 | 3619 | 103 | |
AK159283.1 | - | 47 - 48 | 3615 | 103 | ||
AK159285.1 | - | 47 - 48 | 3619 | 103 | ||
BC003198.1 | - | 10 - 11 | 907 | 103 | ||
BC042642.1 | - | 40 - 41 | 3167 | 103 | ||
BC050014.1 | - | 47 - 48 | 3597 | 4225038 | 103 | |
BC059281.1 | - | 40 - 41 | 2923 | 3992363 | 103 | |
U08020.1 | - | 47 - 48 | 3498 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |