Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eya2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271962.1 | NP_001258891.1 | 5 - 6 | 466 | 62 | |
NM_001271963.1 | NP_001258892.1 | 5 - 6 | 642 | 62 | ||
NM_010165.3 | NP_034295.1 | 5 - 6 | 643 | 62 | ||
GenBank mRNA | AK141765.1 | - | 5 - 6 | 642 | 62 | |
AY212512.1 | - | 4 - 5 | 403 | 62 | ||
BC003755.1 | - | 5 - 6 | 643 | 3497245 | 62 | |
CF724561.1 | - | 4 - 5 | 342 | 62 | ||
U71208.1 | - | 5 - 6 | 468 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |