Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159487.1 | NP_001152959.1 | 4 - 5 | 741 | 96 | |
NM_011255.3 | NP_035385.1 | 4 - 5 | 428 | 96 | ||
GenBank mRNA | AK004839.1 | - | 4 - 5 | 426 | 96 | |
AK008765.1 | - | 4 - 5 | 426 | 96 | ||
AK169338.1 | - | 3 - 4 | 275 | 96 | ||
BC031809.1 | - | 4 - 5 | 406 | 4193984 | 96 | |
BC093529.1 | - | 4 - 5 | 668 | 5135440 | 96 | |
M74527.1 | - | 3 - 4 | 356 | 96 | ||
U63146.1 | - | 3 - 4 | 356 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |