Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011319.3 | NP_035449.1 | 10 - 11 | 1460 | 73 | |
GenBank mRNA | AK082793.1 | - | 10 - 11 | 1371 | 73 | |
BC031932.1 | - | 10 - 11 | 1350 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |