Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001141948.1 | NP_001135420.1 | 6 - 7 | 938 | 96 | |
NM_001141949.1 | NP_001135421.1 | 7 - 8 | 978 | 96 | ||
NM_019401.2 | NP_062274.1 | 6 - 7 | 873 | 96 | ||
GenBank mRNA | AF019249.1 | - | 6 - 7 | 781 | 96 | |
AK146577.1 | - | 7 - 8 | 775 | 96 | ||
BC002019.1 | - | 6 - 7 | 733 | 3590545 | 96 | |
CT010361.1 | - | 5 - 6 | 632 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |