Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001093752.1 | NP_001087221.1 | 8 - 9 | 917 | 120 | |
NM_001093753.1 | NP_001087222.1 | 7 - 8 | 899 | 120 | ||
GenBank mRNA | AK011472.1 | - | 8 - 9 | 855 | 120 | |
AK029394.1 | - | 7 - 8 | 862 | 120 | ||
AK081511.1 | - | 5 - 6 | 724 | 120 | ||
AK146719.1 | - | 7 - 8 | 871 | 120 | ||
BC002111.1 | - | 7 - 8 | 870 | 120 | ||
BC021809.1 | - | 7 - 8 | 854 | 120 | ||
BC037053.1 | - | 8 - 9 | 840 | 120 | ||
BC038928.1 | - | 8 - 9 | 876 | 120 | ||
BC055763.1 | - | 9 - 10 | 804 | 120 | ||
BC069945.1 | - | 7 - 8 | 849 | 120 | ||
BC138145.1 | - | 5 - 6 | 652 | 8861164 | 120 | |
BC145162.1 | - | 5 - 6 | 652 | 9053713 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |