Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027642.2 | NP_081918.1 | 1 - 2 | 222 | 152 | |
GenBank mRNA | AK016480.1 | - | 1 - 2 | 223 | 152 | |
AK038320.1 | - | 1 - 2 | 142 | 152 | ||
AK122560.1 | - | 1 - 2 | 152 | 152 | ||
AK141804.1 | - | 1 - 2 | 165 | 152 | ||
AK157045.1 | - | 1 - 2 | 223 | 152 | ||
AK163345.1 | - | 1 - 2 | 178 | 152 | ||
BC043127.1 | - | 1 - 2 | 170 | 6410361 | 152 | |
BC055330.1 | - | 1 - 2 | 165 | 6406483 | 152 | |
BC057374.1 | - | 1 - 2 | 128 | 6826091 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |