Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300017825.mm.1 |
NM_028188.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017825.mm.1 |
NM_028188.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017825.mm.1 |
TC0300002333.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017828.mm.1 |
NM_001083807.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017828.mm.1 |
NM_001083807.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017828.mm.1 |
NM_028188.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017828.mm.1 |
NM_028188.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017828.mm.1 |
TC0300002333.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017824.mm.1 |
NM_028188.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017824.mm.1 |
NM_028188.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017824.mm.1 |
TC0300002333.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017826.mm.1 |
NM_001083807.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017826.mm.1 |
NM_001083807.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017826.mm.1 |
NM_028188.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017826.mm.1 |
NM_028188.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017826.mm.1 |
TC0300002333.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017827.mm.1 |
NM_001083807.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017827.mm.1 |
NM_001083807.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017827.mm.1 |
NM_028188.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017827.mm.1 |
NM_028188.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017827.mm.1 |
TC0300002333.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017822.mm.1 |
NM_001083807.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017822.mm.1 |
NM_001083807.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017822.mm.1 |
NM_001083808.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017822.mm.1 |
NM_001083808.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017822.mm.1 |
NM_028188.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017822.mm.1 |
NM_028188.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017822.mm.1 |
TC0300002333.mm.1 |
MTA TC |