Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171007.1 | NP_001164478.1 | 3 - 4 | 591 | 69 | |
NM_172729.3 | NP_766317.1 | 3 - 4 | 842 | 69 | ||
GenBank mRNA | AK089662.1 | - | 3 - 4 | 592 | 69 | |
AK137585.1 | - | 3 - 4 | 847 | 69 | ||
AY160222.1 | - | 2 - 3 | 377 | 69 | ||
BC042670.1 | - | 3 - 4 | 669 | 3597911 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |