Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Raver2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183024.1 | NP_898845.1 | 1 - 2 | 288 | 113 | |
GenBank mRNA | AK040404.1 | - | 1 - 2 | 344 | 113 | |
BC055329.1 | - | 1 - 2 | 288 | 6835616 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |