Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tssk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183099.2 | NP_898922.2 | 4 - 5 | 827 | 56 | |
GenBank mRNA | AK007032.1 | - | 4 - 5 | 829 | 56 | |
BC114543.1 | - | 4 - 5 | 631 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |