Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183180.2 | NP_899003.1 | 6 - 7 | 721 | 63 | |
GenBank mRNA | AK032765.1 | - | 5 - 6 | 632 | 63 | |
BC052169.1 | - | 6 - 7 | 684 | 6335221 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |