Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183294.2 | NP_899117.1 | 3 - 4 | 464 | 74 | |
GenBank mRNA | AK016781.1 | - | 3 - 4 | 465 | 74 | |
BC115909.1 | - | 2 - 3 | 412 | 40105122 | 74 | |
BC118055.1 | - | 2 - 3 | 413 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |