Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183427.4 | NP_906272.1 | 1 - 2 | 183 | 103 | |
GenBank mRNA | AK135134.1 | - | 1 - 2 | 328 | 102 | |
AK139335.1 | - | 1 - 2 | 596 | 103 | ||
BC056342.1 | - | 1 - 2 | 183 | 5708257 | 103 | |
BC068987.1 | - | 1 - 2 | 183 | 5708257 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |