Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK034856.1 | - | 10 - 11 | 1326 | 60 | |
AK040344.1 | - | 8 - 9 | 1104 | 60 | ||
AK052103.1 | - | 8 - 9 | 1054 | 60 | ||
AK085336.1 | - | 9 - 10 | 1200 | 60 | ||
AK085362.1 | - | 10 - 11 | 1365 | 60 | ||
AK086462.1 | - | 10 - 11 | 1358 | 60 | ||
BC054120.1 | - | 10 - 11 | 1311 | 6506658 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |