Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ss18l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178750.5 | NP_848865.4 | 8 - 9 | 1253 | 79 | |
GenBank mRNA | AK038669.1 | - | 8 - 9 | 948 | 79 | |
AK054560.1 | - | 8 - 9 | 968 | 79 | ||
AK133944.1 | - | 8 - 9 | 1190 | 79 | ||
AK173004.1 | - | 8 - 9 | 1006 | 79 | ||
BC053087.1 | - | 8 - 9 | 1077 | 5708900 | 79 | |
BC054730.1 | - | 8 - 9 | 1227 | 5708860 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |