Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291190.1 | NP_001278119.1 | 8 - 9 | 991 | 125 | |
NM_177710.4 | NP_808378.2 | 9 - 10 | 855 | 125 | ||
GenBank mRNA | AB099288.1 | - | 9 - 10 | 855 | 125 | |
AK155232.1 | - | 8 - 9 | 982 | 125 | ||
AK170142.1 | - | 8 - 9 | 988 | 125 | ||
BC141392.1 | - | 9 - 10 | 849 | 9056005 | 125 | |
BC141393.1 | - | 9 - 10 | 849 | 9056006 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |