Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mef2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291191.1 | NP_001278120.1 | 8 - 9 | 1222 | 62 | |
NM_001291192.1 | NP_001278121.1 | 8 - 9 | 1222 | 62 | ||
NM_001291195.1 | NP_001278124.1 | 8 - 9 | 1196 | 62 | ||
NM_001291196.1 | NP_001278125.1 | 9 - 10 | 1343 | 62 | ||
GenBank mRNA | AK040227.1 | - | 4 - 5 | 1118 | 62 | |
AK052385.1 | - | 9 - 10 | 1344 | 62 | ||
AK132678.1 | - | 8 - 9 | 1078 | 62 | ||
AK153751.1 | - | 8 - 9 | 1223 | 62 | ||
BC096598.1 | - | 8 - 9 | 1164 | 4979487 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |