Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apbb1ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019456.2 | NP_062329.2 | 6 - 7 | 896 | 108 | |
GenBank mRNA | AF020313.1 | - | 3 - 4 | 502 | 108 | |
AK021331.1 | - | 7 - 8 | 775 | 108 | ||
AK040084.1 | - | 6 - 7 | 832 | 108 | ||
AK041552.1 | - | 6 - 7 | 899 | 108 | ||
AK042467.1 | - | 6 - 7 | 834 | 108 | ||
AK079736.1 | - | 6 - 7 | 1130 | 108 | ||
BC003991.1 | - | 3 - 4 | 547 | 108 | ||
BC023110.1 | - | 6 - 7 | 785 | 3707478 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |