Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1978511 |
AY989883.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1978511 |
BC120554.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1978511 |
BP762043.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1978511 |
NM_001201460.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1978511 |
NM_001198561.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1978511 |
AY989883.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1978511 |
BC120554.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1978511 |
BP762043.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1978511 |
NM_001201460.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1978511 |
NM_001198561.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
AY989883.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
BC120554.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
NM_001201460.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
NM_010394.4 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
X05389.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
AY989883.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
BC120554.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
NM_001201460.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
NM_010394.4 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
X05389.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
AY989883.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
BC120554.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
NM_001201460.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
NM_010394.4 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
X05389.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
AY989883.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
BC120554.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
NM_001201460.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
NM_010394.4 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
X05389.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
AY989883.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
BC120554.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
NM_001201460.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
NM_010394.4 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
X05389.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
AY989883.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
BC120554.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
NM_001201460.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
NM_010394.4 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
X05389.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
AY989883.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
BC120554.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
NM_001201460.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
NM_010394.4 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
X05389.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
AY989883.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
BC120554.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
NM_001201460.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
NM_010394.4 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
X05389.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
AY989883.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
BC120554.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
NM_001201460.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
NM_010394.4 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
X05389.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
AY989883.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
BC120554.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
NM_001201460.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
NM_010394.4 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P262079 |
X05389.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
AY989883.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
BC120554.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
NM_001201460.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
NM_010394.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
X05389.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
AY989883.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
BC120554.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
NM_001201460.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
NM_010394.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
X05389.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
AY989883.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
BC120554.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
NM_001201460.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
NM_010394.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
X05389.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
AY989883.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
BC120554.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
NM_001201460.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
NM_010394.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
X05389.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
AY989883.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
BC120554.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
NM_001201460.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
NM_010394.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
X05389.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
AY989883.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
BC120554.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
NM_001201460.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
NM_010394.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
X05389.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
AY989883.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
BC120554.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
NM_001201460.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
NM_010394.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
X05389.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
AY989883.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
BC120554.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
NM_001201460.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
NM_010394.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
X05389.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
AY989883.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
BC120554.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
NM_001201460.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
NM_010394.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
X05389.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
AY989883.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
BC120554.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
NM_001201460.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
NM_010394.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P262079 |
X05389.1 |
mouse_transcriptome |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31159173 |
- |
- |