Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900023201.mm.1 |
NM_001284328.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023201.mm.1 |
NM_019796.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023201.mm.1 |
TC0900003342.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023198.mm.1 |
NM_001284328.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023198.mm.1 |
NM_019796.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023198.mm.1 |
TC0900003342.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023195.mm.1 |
NM_001284328.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023195.mm.1 |
NM_019796.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023195.mm.1 |
TC0900003342.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023191.mm.1 |
NM_001284328.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023191.mm.1 |
NM_019796.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023191.mm.1 |
TC0900003342.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023200.mm.1 |
NM_001284328.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023200.mm.1 |
NM_019796.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023200.mm.1 |
TC0900003342.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900028054.mm.1 |
NM_001284328.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900028054.mm.1 |
NM_019796.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900028054.mm.1 |
TC0900003342.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023203.mm.1 |
NM_001284328.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023203.mm.1 |
NM_019796.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023203.mm.1 |
TC0900003342.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023197.mm.1 |
NM_001284328.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023197.mm.1 |
NM_019796.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023197.mm.1 |
TC0900003342.mm.1 |
MTA TC |