Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agbl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048192.2 | NP_001041657.1 | 9 - 10 | 1742 | 65 | |
NM_001294262.1 | NP_001281191.1 | 8 - 9 | 1770 | 65 | ||
NM_174849.3 | NP_777274.1 | 8 - 9 | 1659 | 65 | ||
NR_122116.1 | 8 - 9 | 1726 | 65 | |||
NR_122117.1 | 8 - 9 | 1726 | 65 | |||
GenBank mRNA | AK029676.1 | - | 8 - 9 | 1660 | 65 | |
AK034894.1 | - | 8 - 9 | 1748 | 65 | ||
AK035680.1 | - | 6 - 7 | 1227 | 65 | ||
BC057349.1 | - | 8 - 9 | 1660 | 6813901 | 65 | |
DQ867034.1 | - | 8 - 9 | 1655 | 65 | ||
DQ867035.1 | - | 8 - 9 | 1655 | 65 | ||
DQ867036.1 | - | 9 - 10 | 1742 | 65 | ||
DQ867037.1 | - | 8 - 9 | 1655 | 65 | ||
DQ867038.1 | - | 8 - 9 | 1655 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |