Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028399.1 | NP_082675.1 | 5 - 6 | 554 | 89 | |
GenBank mRNA | AK036772.1 | - | 502 | 89 | ||
AK049863.1 | - | 5 - 6 | 499 | 89 | ||
AK152399.1 | - | 5 - 6 | 502 | 89 | ||
AK154439.1 | - | 5 - 6 | 512 | 89 | ||
AK156231.1 | - | 523 | 89 | |||
BC054122.1 | - | 5 - 6 | 554 | 6509024 | 89 | |
JX965957.1 | - | 5 - 6 | 490 | 89 | ||
JX965958.1 | - | 5 - 6 | 490 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |