Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146087.2 | NP_666199.1 | 4 - 5 | 617 | 111 | |
GenBank mRNA | AK076070.1 | - | 4 - 5 | 884 | 111 | |
AK076264.1 | - | 4 - 5 | 878 | 111 | ||
AK078398.1 | - | 4 - 5 | 866 | 111 | ||
AK141421.1 | - | 4 - 5 | 872 | 111 | ||
AK146873.1 | - | 4 - 5 | 887 | 111 | ||
AK147165.1 | - | 4 - 5 | 862 | 111 | ||
AK152387.1 | - | 4 - 5 | 862 | 111 | ||
AK152793.1 | - | 4 - 5 | 882 | 111 | ||
AK159939.1 | - | 1 - 2 | 86 | 111 | ||
AK166318.1 | - | 4 - 5 | 877 | 111 | ||
AK166429.1 | - | 4 - 5 | 859 | 111 | ||
AK167550.1 | - | 4 - 5 | 885 | 111 | ||
BC019740.1 | - | 4 - 5 | 617 | 3487913 | 111 | |
BC025439.1 | - | 4 - 5 | 564 | 5038433 | 111 | |
BC048081.1 | - | 4 - 5 | 650 | 6309504 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus 1700 Probe spans exons |