Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290651.1 | NP_001277580.1 | 21 - 22 | 2609 | 164 | |
NM_001290652.1 | NP_001277581.1 | 19 - 20 | 2429 | 164 | ||
GenBank mRNA | AF285577.1 | - | 18 - 19 | 2015 | 164 | |
AK016533.1 | - | 18 - 19 | 1978 | 164 | ||
AK031490.1 | - | 21 - 22 | 2590 | 164 | ||
AK040521.1 | - | 19 - 20 | 2431 | 164 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |