Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcdc2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195617.1 | NP_001182546.1 | 6 - 7 | 1025 | 64 | |
NM_177577.3 | NP_808245.2 | 6 - 7 | 1025 | 64 | ||
GenBank mRNA | AK220370.1 | - | 6 - 7 | 798 | 64 | |
BC045136.1 | - | 6 - 7 | 1025 | 6306251 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |