Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007462.3 | NP_031488.2 | 7 - 8 | 1031 | 79 | |
GenBank mRNA | AK045053.1 | - | 7 - 8 | 1002 | 79 | |
AK046280.1 | - | 7 - 8 | 1021 | 79 | ||
AK080907.1 | - | 7 - 8 | 1031 | 79 | ||
AK082724.1 | - | 7 - 8 | 777 | 79 | ||
BC141141.1 | - | 6 - 7 | 744 | 9055753 | 79 | |
M88127.1 | - | 6 - 7 | 720 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |