Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026853.1 | NP_081129.1 | 3 - 4 | 306 | 63 | |
GenBank mRNA | AF398972.1 | - | 3 - 4 | 420 | 63 | |
AK002412.1 | - | 3 - 4 | 393 | 63 | ||
AK004396.1 | - | 3 - 4 | 306 | 63 | ||
AK005404.1 | - | 3 - 4 | 420 | 63 | ||
AK146664.1 | - | 3 - 4 | 420 | 63 | ||
BC064047.1 | - | 3 - 4 | 302 | 864428 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |