Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019467.2 | NP_062340.1 | 6 - 7 | 512 | 111 | |
GenBank mRNA | AB013745.1 | - | 4 - 5 | 239 | 111 | |
AF074959.1 | - | 4 - 5 | 250 | 111 | ||
AK006184.1 | - | 6 - 7 | 512 | 111 | ||
AK006562.1 | - | 6 - 7 | 513 | 111 | ||
BC021539.1 | - | 4 - 5 | 380 | 5346103 | 111 | |
CT010265.1 | - | 4 - 5 | 198 | 111 | ||
CT010385.1 | - | 4 - 5 | 198 | 111 | ||
D86382.1 | - | 4 - 5 | 275 | 111 | ||
EF101555.1 | - | 3 - 4 | 150 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |