Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikzf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011772.2 | NP_035902.2 | 1 - 2 | 277 | 122 | |
GenBank mRNA | AK089522.1 | - | 1 - 2 | 111 | 122 | |
AK165740.1 | - | 541 | 122 | |||
AK170453.1 | - | 1 - 2 | 129 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |