Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK082940.1 | - | 11 - 12 | 1094 | 76 | |
AK167383.1 | - | 11 - 12 | 1082 | 76 | ||
AK167848.1 | - | 11 - 12 | 1078 | 76 | ||
AY082332.1 | - | 11 - 12 | 1048 | 76 | ||
BC050206.1 | - | 11 - 12 | 1056 | 30009463 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |