Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144908.3 | NP_659157.1 | 2 - 3 | 472 | 82 | |
GenBank mRNA | AF495528.1 | - | 1 - 2 | 344 | 82 | |
AK088014.1 | - | 2 - 3 | 461 | 82 | ||
AK161524.1 | - | 2 - 3 | 382 | 82 | ||
BC011428.1 | - | 2 - 3 | 448 | 2631696 | 82 | |
BC021504.1 | - | 2 - 3 | 473 | 5346678 | 82 | |
BC031150.1 | - | 1 - 2 | 372 | 82 | ||
BC034184.1 | - | 1 - 2 | 372 | 4974216 | 82 | |
BC034185.1 | - | 1 - 2 | 424 | 4974695 | 82 | |
BC036143.1 | - | 1 - 2 | 373 | 4974645 | 82 | |
BC036145.1 | - | 1 - 2 | 366 | 4975474 | 82 | |
Y12435.1 | - | 1 - 2 | 295 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |