Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004365.1 | NP_001004365.1 | 4 - 5 | 401 | 69 | |
GenBank mRNA | AK035798.1 | - | 3 - 4 | 313 | 69 | |
BC053106.1 | - | 3 - 4 | 374 | 69 | ||
BC082279.1 | - | 4 - 5 | 401 | 6332588 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |