Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170555.1 | NP_001164026.1 | 8 - 9 | 830 | 102 | |
NM_001170556.1 | NP_001164027.1 | 8 - 9 | 827 | 102 | ||
NM_145401.2 | NP_663376.2 | 12 - 13 | 1909 | 102 | ||
GenBank mRNA | AK032238.1 | - | 10 - 11 | 1421 | 102 | |
AK034765.1 | - | 6 - 7 | 495 | 102 | ||
AK154296.1 | - | 12 - 13 | 1910 | 102 | ||
AK165834.1 | - | 8 - 9 | 769 | 102 | ||
AY348864.1 | - | 8 - 9 | 760 | 102 | ||
BC015283.1 | - | 12 - 13 | 1697 | 4238045 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |