Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011203.2 | NP_035333.2 | 6 - 7 | 539 | 85 | |
GenBank mRNA | AK154107.1 | - | 6 - 7 | 631 | 85 | |
BC051980.1 | - | 6 - 7 | 539 | 6402545 | 85 | |
X63440.1 | - | 6 - 7 | 540 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |