Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300010507.mm.1 |
NM_001271378.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010507.mm.1 |
NM_001271379.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010507.mm.1 |
NM_011167.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010507.mm.1 |
NR_073166.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010507.mm.1 |
TC1300001690.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300010504.mm.1 |
NM_001271378.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010504.mm.1 |
NM_001271379.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010504.mm.1 |
NM_011167.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010504.mm.1 |
NR_073166.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010504.mm.1 |
TC1300001690.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300010502.mm.1 |
NM_001271378.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010502.mm.1 |
NM_001271379.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010502.mm.1 |
NM_011167.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010502.mm.1 |
TC1300001690.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300010505.mm.1 |
NM_001271378.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010505.mm.1 |
NM_001271379.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010505.mm.1 |
NM_011167.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010505.mm.1 |
NR_073166.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300010505.mm.1 |
TC1300001690.mm.1 |
MTA TC |