Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc13a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019481.2 | NP_062354.2 | 2 - 3 | 260 | 95 | |
GenBank mRNA | AF199365.1 | - | 2 - 3 | 260 | 95 | |
AF199366.1 | - | 2 - 3 | 260 | 95 | ||
AK019876.1 | - | 2 - 3 | 259 | 95 | ||
AK128899.1 | - | 2 - 3 | 260 | 95 | ||
BC040789.1 | - | 2 - 3 | 234 | 95 | ||
BC049981.1 | - | 2 - 3 | 242 | 5065026 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |