Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138587.4 | NP_613053.3 | 8 - 9 | 588 | 91 | |
GenBank mRNA | AK004059.1 | - | 1 - 2 | 83 | 91 | |
AK154498.1 | - | 8 - 9 | 618 | 91 | ||
AK170887.1 | - | 8 - 9 | 637 | 91 | ||
AY424275.1 | - | 8 - 9 | 588 | 91 | ||
BC009086.1 | - | 8 - 9 | 588 | 2811008 | 91 | |
BC069880.1 | - | 7 - 8 | 714 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |