Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291320.1 | NP_001278249.1 | 2 - 3 | 316 | 60 | |
NM_133709.3 | NP_598470.3 | 2 - 3 | 358 | 60 | ||
GenBank mRNA | AF338222.1 | - | 2 - 3 | 262 | 60 | |
AK007577.1 | - | 2 - 3 | 320 | 60 | ||
BC019399.1 | - | 2 - 3 | 323 | 4012143 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |