Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021885.4 | NP_068685.1 | 1 - 2 | 254 | 63 | |
GenBank mRNA | AK147700.1 | - | 1 - 2 | 309 | 63 | |
AK165720.1 | - | 1 - 2 | 255 | 63 | ||
BC096554.1 | - | 1 - 2 | 235 | 30616697 | 63 | |
U52433.1 | - | 1 - 2 | 242 | 63 | ||
U54643.1 | - | 1 - 2 | 172 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |