Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285959.1 | NP_001272888.1 | 2 - 3 | 194 | 137 | |
NM_001285960.1 | NP_001272889.1 | 2 - 3 | 327 | 137 | ||
NM_029610.3 | NP_083886.1 | 2 - 3 | 578 | 137 | ||
GenBank mRNA | AK009135.1 | - | 2 - 3 | 581 | 137 | |
AK015083.1 | - | 3 - 4 | 386 | 137 | ||
AK157931.1 | - | 196 | 137 | |||
BC011339.1 | - | 2 - 3 | 368 | 3978191 | 137 | |
BY752674.1 | - | 2 - 3 | 196 | 137 | ||
DV043140.1 | - | 1 - 2 | 352 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |