Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010689.2 | NP_034819.1 | 2 - 3 | 428 | 63 | |
GenBank mRNA | AF036907.1 | - | 2 - 3 | 428 | 63 | |
AK088763.1 | - | 2 - 3 | 337 | 63 | ||
AK134003.1 | - | 2 - 3 | 380 | 63 | ||
BC013337.1 | - | 2 - 3 | 405 | 4191983 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |