Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162924.1 | NP_001156396.1 | 5 - 6 | 1237 | 92 | |
NM_019762.2 | NP_062736.2 | 4 - 5 | 1168 | 92 | ||
GenBank mRNA | AF136719.1 | - | 4 - 5 | 1159 | 92 | |
BC020472.1 | - | 4 - 5 | 1140 | 92 | ||
BC032045.1 | - | 4 - 5 | 1128 | 92 | ||
BC090668.1 | - | 5 - 6 | 1237 | 30696296 | 92 | |
BC106141.1 | - | 4 - 5 | 1133 | 92 | ||
BC139775.1 | - | 4 - 5 | 1124 | 40111915 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |